Tetradecyltrimethylammoniumchlorid ≥98%, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Myristyltrimethylammoniumchlorid, Tetradoniumchlorid, TTAC
87212-5G
SIAL87212-5GEA
64.4
EUR
SIAL87212-5G
Tetradecyltrimethylammoniumchlorid ≥98%, Sigma-Aldrich®
Tetradecyltrimethylammoniumchlorid
Formel:
C₁₇H₃₈ClN Molecular Weight: 291.95 g/mol |
MDL:
MFCD00059968 CAS-Nummer: 4574-04-3 EINECS: 224-958-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...