N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N,N,N-Tetrakis(2-hydroxyethyl)ethylenediamine

87600-100ML
SIAL87600-100MLEA 39.1 EUR
SIAL87600-100ML
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylendiamin, Sigma-Aldrich®
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylendiamin
Formel: C₁₀H₂₄N₂O₄
Molecular Weight: 236.3086 g/mol
MDL: MFCD00002854
CAS-Nummer: 140-07-8
EINECS: 205-396-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR