Tetrakis(trimethylsilyl)silan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: TMSS

M-Clarity™ Qualitätsstufe = MQ100

87642-5G
SIAL87642-5GEA 137 EUR
SIAL87642-5G
Tetrakis(trimethylsilyl)silan, Sigma-Aldrich®
Tetrakis(trimethylsilyl)silan
Formel: C₁₂H₃₆Si₅
Molecular Weight: 320,842 g/mol
MDL: MFCD00054859
CAS-Nummer: 4098-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR