Tetramethylammoniumhydrogensulfat Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetramethylammonium hydrogensulphate hydrate

87726-50G
SIAL87726-50GEA 327 EUR
SIAL87726-50G
Tetramethylammoniumhydrogensulfat Monohydrat, Sigma-Aldrich®
Tetramethylammoniumhydrogensulfat Monohydrat
Formel: C₄H₁₃NO₄S·1H₂O
Molecular Weight: 189,23 g/mol
MDL: MFCD00150356
CAS-Nummer: 103812-00-6
EINECS: 279-490-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR