3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydrochloride TMB, TMB-HCl

87750-1G 87750-25G 87750-5G
SIAL87750-1GEA 89.2 EUR
SIAL87750-1G SIAL87750-25G SIAL87750-5G
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel: C₁₆H₂₀N₂·2HCl·xH₂O
Molecular Weight: 313.27 g/mol (anhy)
MDL: MFCD00150104
CAS-Nummer: 207738-08-7
EINECS: 264-769-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR