DSPE-PEG(2000) Biotin, Pulver, Avanti Research™

Lieferant: Merck

Synonyme: 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(polyethylene glycol)-2000] (ammonium salt)

880129P-50MG
SIAL880129P-50MGEA 0 EUR
SIAL880129P-50MG
DSPE-PEG(2000) Biotin, Pulver, Avanti Research™
DSPE-PEG(2000) Biotin

DSPE-PEG(2000) biotin is functionalised polyethylene glycol (PEG) conjugated lipid.


  • DSPE-PEG(2000) biotin has been used as a component of liposomes, to prepare lipid microbubbles
  • Used in the control experiments, to test the influence of 2,4-dinitrophenyl (DNP)-PEG2000-PE conformation on binding affinity
  • Used for capsule surface modification
  • Added to the labelling reaction to facilitate immobilisation of the samples on the streptavidin-coated microfluidic sample chambers


Avanti Research™ - A Croda Brand

Lagertemperatur: Gefrierschrank
CAS-Nummer: 385437-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR