DSPE-PEG-DBCO, M.W. 2000, Avanti Research™

Lieferant: Merck

880229P-50MG
SIAL880229P-50MGEA 0 EUR
SIAL880229P-50MG
DSPE-PEG-DBCO, M.W. 2000, Avanti Research™
DSPE-PEG-DBCO

DSPE-PEG(2000)-DBCO [1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[dibenzocyclooctyl(polyethylene glycol)-2000] (ammonium salt)] is a functionalised polyethylene glycol (PEG) lipid conjugate.


DSPE-PEG(2000)-DBCO has been used in the preparation of peptide conjugated fluorescent nanoparticle by modified thin-film hydration technique. It has also been used for homeotropic anchoring of the mesogen molecules.


Avanti Research™ - A Croda Brand

Formel: (C₂H₄O)nC₆₃H₁₀₀N₃O₁₂P.H₃N
Lagertemperatur: Gefrierschrank
CAS-Nummer: 2052955-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR