Titanium(IV) oxide sulphate sulphuric acid hydrate-Lösung (27 - 31% H₂SO₄ basis), Millipore®

Lieferant: Merck
Gefahr

89532-1L
SIAL89532-1LEA 123 EUR
SIAL89532-1L
Titanium(IV) oxide sulphate sulphuric acid hydrate-Lösung (27 - 31% H₂SO₄ basis), Millipore®
Titanium(IV) oxide sulphate sulphuric acid hydrate

For the detection of H₂O₂ and ether peroxides.

Formel: TiOSO₄·xH₂SO₄·yH₂O
MDL: MFCD00150565
CAS-Nummer: 123334-00-9

Bestellen

Spezifikation Testergebnisse

Density 1,32 g/ml at 20 °C
Assay 27 - 31% H₂SO₄ basis ~5% Ti basis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR