(R)-3-(Boc-amino)-3-(4-methylphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-(p-tolyl)propanoic acid

90297-500MG-F
SIAL90297-500MG-FEA 354 EUR
SIAL90297-500MG-F
(R)-3-(Boc-amino)-3-(4-methylphenyl)propionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-(4-methylphenyl)propionsäure
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,3315 g/mol
MDL: MFCD03427935
CAS-Nummer: 479064-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR