(S)-3-(Boc-amino)-3-(4-methylphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(p-tolyl)propanoic acid

90298-500MG-F
SIAL90298-500MG-FEA 340 EUR
SIAL90298-500MG-F
(S)-3-(Boc-amino)-3-(4-methylphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(4-methylphenyl)propionsäure
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,3315 g/mol
MDL: MFCD03427897
CAS-Nummer: 479064-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR