Triethylenglycolmonomethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: mPEG3-Alcohol, 2-(2-(2-Methoxyethoxy)ethoxy)ethanol, Methyl triglycol, m-PEG3-OH

MQ200

90450-1L 90450-250ML
SIAL90450-1LEA 116 EUR
SIAL90450-1L SIAL90450-250ML
Triethylenglycolmonomethylether, Sigma-Aldrich®
Triethylenglycolmonomethylether
Formel: C₇H₁₆O₄
Molecular Weight: 164.2 g/mol
MDL: MFCD00002875
CAS-Nummer: 112-35-6
EINECS: 203-962-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR