DL-α-Tocopherol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-α-Tocopherol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, Vitamin E

M-Clarity™ Qualitätsstufe = MQ200

90669-50G-F
SIAL90669-50G-FEA 141 EUR
SIAL90669-50G-F
DL-α-Tocopherol, Sigma-Aldrich®
DL-α-Tocopherol
Formel: C₂₉H₅₀O₂
Molecular Weight: 430,71 g/mol
MDL: MFCD00072051
CAS-Nummer: 10191-41-0
EINECS: 233-466-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR