Methyltributylammoniumbromid, Sigma-Aldrich®

Lieferant: Merck

90802-100G
SIAL90802-100GEA 337 EUR
SIAL90802-100G
Methyltributylammoniumbromid, Sigma-Aldrich®
Methyltributylammonium bromide
Formel: C₉H₂₂BrN
Molecular Weight: 280.29 g/mol
MDL: MFCD00043201
CAS-Nummer: 37026-88-3
EINECS: 253-313-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR