RajHans ChromoSelect medium, modified suitable for microbiology, NutriSelect® Plus, Millipore®
Lieferant: Merck
90918-250G
SIAL90918-250GEA 358 EUR
SIAL90918-250G
RajHans ChromoSelect medium, modified suitable for microbiology, NutriSelect® Plus, Millipore®
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
RajHans ChromoSelect Medium is recommended for identification and differentiation of Salmonella species from the members of Enterobacteriaceae, especially Proteus species.
  • Final pH: 7,3 ±0,2 (25°C)
  • Appearance: Pinkish yellow to beige coloured