1,3,5-Trichlor-2-nitrobenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Nitro-2,4,6-trichlorbenzol, 2,4,6-Trichlornitrobenzol

91315-25G
SIAL91315-25GEA 81.2 EUR
SIAL91315-25G
1,3,5-Trichlor-2-nitrobenzol, Sigma-Aldrich®
1,3,5-Trichlor-2-nitrobenzol
Formel: C₆H₂Cl₃NO₂
Molecular Weight: 226,45 g/mol
Schmelzpunkt: 71 °C
MDL: MFCD00014690
CAS-Nummer: 18708-70-8
EINECS: 242-518-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR