1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluor-12-ioddodecan, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1H,1H,2H,2H-Perfluorododecyl iodide, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Henicosafluoro-12-iodododecane, 1H,1H,2H,2H-Perfluoro-1-iodododecane

91987-2.5G-F
SIAL91987-2.5G-FEA 79 EUR
SIAL91987-2.5G-F
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluor-12-ioddodecan, Sigma-Aldrich®
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluor-12-ioddodecan
Formel: C₁₂H₄F₂₁I
Molecular Weight: 674,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039412
CAS-Nummer: 2043-54-1
EINECS: 218-054-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR