Copper indium sulphide zinc sulphide (CuInS2/ZnS) 1 mg/ml in Toluol, Kern-Schale-Quantenpunkte fluorescence λem 650 nm, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

924326-1MG
SIAL924326-1MGEA 297 EUR
SIAL924326-1MG
Copper indium sulphide zinc sulphide (CuInS2/ZnS) 1 mg/ml in Toluol, Kern-Schale-Quantenpunkte fluorescence λem 650 nm, Sigma-Aldrich®
Copper indium sulphide zinc sulphide (CuInS2/ZnS)

Copper indium sulfide zinc sulfide (CuInS2/ZnS) core/shell quantum dots are a heavy metal-free, high-performance alternative to cadmium-based quantum dots. CuInS2 QDs, as the I–III–VI ternary semiconductor nanocrystals contain two cations (Cu+ and In3+) and one anion (S2–). These QDs have unique photophysical properties; that make them superior to molecular dyes. Advantages of QDs include higher photostability, broad absorption spectrum, high extinction coefficients, and tunable optical and electronic properties.


CuInS2/ZnS quantum dots possess brilliant quantum yield and excellent long-term stability, and these non-toxic quantum dots lack the environmental hazards of cadmium-based quantum dots.


Bitte beachten Do not freeze.

Flammpunkt: 4,4 °C (39.9 °F)
Lagertemperatur: Kühlschrank
UN: 1294
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

fluorescence FWHM <125 nm - λem 650 nm, quantum yield ≥40%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR