(S)-3-(Boc-amino)-3-(4-bromphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

93123-500MG-F
SIAL93123-500MG-FEA 319 EUR
SIAL93123-500MG-F
(S)-3-(Boc-amino)-3-(4-bromphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(4-bromphenyl)propionsäure
Formel: C₁₄H₁₈BrNO₄
Molecular Weight: 344,2010 g/mol
MDL: MFCD03419292
CAS-Nummer: 261165-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR