Tris-EDTA buffer solution, pH 8,0, for molecular biology, BioUltra, Sigma-Aldrich®
Lieferant: Merck
93283-100ML
93283-500ML
SIAL93283-100MLEA
51.9
EUR
SIAL93283-100ML
SIAL93283-500ML
Tris-EDTA buffer solution, pH 8,0, for molecular biology, BioUltra, Sigma-Aldrich®
Puffer
Biologische Puffer
This buffer is 10 mM Tris-HCl and 1 mM disodium EDTA.
- UV absorption - λ: 260 nm; Amax: 0,04 and λ: 280 nm Amax: 0,01
- Contains traces of metal cations
Trizma is used in the formulation of buffer solutions in the pH range between 7,5 and 8,5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8,0 may also be used as a washing buffer.