Tropin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

93550-10G
SIAL93550-10GEA 53 EUR
SIAL93550-10G
Tropin, Sigma-Aldrich®
Tropin
Formel: C₈H₁₅NO
Molecular Weight: 141.21 g/mol
MDL: MFCD00180667
CAS-Nummer: 120-29-6
EINECS: 204-384-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR