Atto 647N-biotin, BioReagent

Lieferant: Merck

93606-1MG
SIAL93606-1MGEA 490 EUR
SIAL93606-1MG
Atto 647N-biotin, BioReagent
Proteinkonjugate und Markierungen

Designed for high sensitivity applications, including single molecule detection. These labels have rigid structures that do not show any cis-trans-isomerisation and display exceptional intensity with minimal spectral shift on conjugation.


  • Assay: ≥95% (HPCE)
  • Fluorescence: λex 644 nm; λem 660 nm in 0,1 M phosphate pH 7,0
  • Suitable for fluorescence

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR