L(-)-Tryptophan, Sigma-Aldrich®
Lieferant: Merck
93659-10G
93659-250G
93659-50G
SIAL93659-10GEA
80.3
EUR
SIAL93659-10G
SIAL93659-250G
SIAL93659-50G
L(-)-Tryptophan, Sigma-Aldrich®
L(-)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Schmelzpunkt: 290 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064340 CAS-Nummer: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...