Tween® 20 (Polysorbat), Sigma-Aldrich®
Lieferant: Merck
93773-1KG
SIAL93773-1KGEA
99
EUR
SIAL93773-1KG
Tween® 20 (Polysorbat), Sigma-Aldrich®
Tween® 20 (Polysorbat)
Formel:
C₅₈H₁₁₄O₂₆ Dichte: 1,1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00165986 CAS-Nummer: 9005-64-5 EINECS: 500-018-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...