DL-o-Tyrosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxyphenylalanin, 3-(o-Hydroxyphenyl)-DL-alanin, 2-Amino-3-(2-hydroxyphenyl)propionsäure

93851-100MG
SIAL93851-100MGEA 134 EUR
SIAL93851-100MG
DL-o-Tyrosin, Sigma-Aldrich®
DL-o-Tyrosin
Formel: C₉H₁₁NO₃
MDL: MFCD00021725
CAS-Nummer: 2370-61-8
EINECS: 219-134-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR