10-Undecen-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Undecin-11-ol

94195-1ML
SIAL94195-1MLEA 78.8 EUR
SIAL94195-1ML
10-Undecen-1-ol, Sigma-Aldrich®
10-Undecin-1-ol
Formel: C₁₁H₂₀O
Molecular Weight: 168,28 g/mol
MDL: MFCD00041675
CAS-Nummer: 2774-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR