(2-Methyl-2-propanyl)lithium 1.6-3.2 mol/l in Heptan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: t-Butyllithium

94439-100ML-F
SIAL94439-100ML-FEA 176 EUR
SIAL94439-100ML-F
(2-Methyl-2-propanyl)lithium 1.6-3.2 mol/l in Heptan, Sigma-Aldrich®
tert-Butyllithium
Formel: C₄H₉Li
Molecular Weight: 64.06 g/mol
MDL: MFCD00008795
CAS-Nummer: 594-19-4
EINECS: 209-831-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR