Vanadyl(IV)acetylacetonat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
94735-50G
SIAL94735-50GEA
47.7
EUR
SIAL94735-50G
Vanadyl(IV)acetylacetonat, Sigma-Aldrich®
Vanadyl(IV)acetylacetonat
Formel:
C₁₀H₁₄O₅V |
MDL:
MFCD00000032 CAS-Nummer: 3153-26-2 EINECS: 221-590-8 UN: 3285 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...