4-Brom-1-butanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-4-hydroxybutan, Tetramethylenbromhydrin

95517-2.5G-F
SIAL95517-2.5G-FEA 108 EUR
SIAL95517-2.5G-F
4-Brom-1-butanol, Sigma-Aldrich®
4-Brom-1-butanol
Formel: BrH₂C(CH₂)₂CH₂OH
Molecular Weight: 153,02 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00015387
CAS-Nummer: 33036-62-3
EINECS: 251-355-5
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR