Polysorbat 60, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
95754-100G-F
SIAL95754-100G-FEA
60.4
EUR
SIAL95754-100G-F
Polysorbat 60, Sigma-Aldrich®
Polysorbat 60
Formel:
C₆₄H₁₂₆O₂₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01780110 CAS-Nummer: 9005-67-8 EINECS: 500-020-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...