N-Acetyl-D-leucin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Acetyle-D-Leucine, N-Ac-D-Leu-OH, Ac-D-Leu-OH, 2-(Acetylamino)-4-methylpentanoic acid

M-Clarity™ Qualitätsstufe = MQ200

A0876-5G
SIALA0876-5GEA 551 EUR
SIALA0876-5G
N-Acetyl-D-leucin, Sigma-Aldrich®
N-Acetyl-D-leucin
Formel: C₈H₁₅NO₃
Molecular Weight: 173,2 g/mol
MDL: MFCD00066069
CAS-Nummer: 19764-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR