N-Acetyl-D-methionin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ac-D-Met-OH, N-Ac-D-Met-OH, (2R)-2-acetamido-4-(methylsulfanyl)butanoic acid, N-Acetyl-D-Methionine

MQ400

A1501-5G
SIALA1501-5GEA 439 EUR
SIALA1501-5G
N-Acetyl-D-methionin, Sigma-Aldrich®
N-Acetyl-D-methionin
Formel: C₇H₁₃NO₃S
Molecular Weight: 191.2 g/mol
MDL: MFCD00069962
CAS-Nummer: 1509-92-8
EINECS: 216-144-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR