Ampicillin Trihydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

M-Clarity™ Qualitätsstufe = MQ200

A1593-25G
SIALA1593-25GEA 462 EUR
SIALA1593-25G
Ampicillin Trihydrat, Sigma-Aldrich®
Ampicillin Trihydrat
Formel: C₁₆H₁₉N₃O₄S·3H₂O
Molecular Weight: 403,45 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00072036
CAS-Nummer: 7177-48-2
EINECS: 200-709-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR