N-Acetylglycin (Ac-Gly-OH), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

A16300-100G A16300-500G A16300-5G
SIALA16300-100GEA 49.6 EUR
SIALA16300-100G SIALA16300-500G SIALA16300-5G
N-Acetylglycin (Ac-Gly-OH), Sigma-Aldrich®
N-Acetylglycin (Ac-Gly-OH)
Formel: C₄H₇NO₃
MDL: MFCD00004275
CAS-Nummer: 543-24-8
EINECS: 208-839-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR