D(+)-α-Acetobromglucose, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose
M-Clarity™ Qualitätsstufe = MQ300
A1750-100G
A1750-10G
A1750-25G
A1750-50G
SIALA1750-100GEA
687
EUR
SIALA1750-100G
SIALA1750-10G
SIALA1750-25G
SIALA1750-50G
D(+)-α-Acetobromglucose, Sigma-Aldrich®
D(+)-α-Acetobromglucose
Formel:
C₁₄H₁₉BrO₉ Molecular Weight: 411.2 g/mol |
MDL:
MFCD00063254 CAS-Nummer: 572-09-8 EINECS: 209-339-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...