6-Azauracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2,4-Triazine-3,5(2H,4H)-dione

A1757-5G
SIALA1757-5GEA 86.5 EUR
SIALA1757-5G
6-Azauracil, Sigma-Aldrich®
6-Azauracil
Formel: C₃H₃N₃O₂
Molecular Weight: 113,08 g/mol
MDL: MFCD00006456
CAS-Nummer: 461-89-2
EINECS: 207-318-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR