N-Acetyl-5-hydroxytryptamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(2-(5-Hydroxy-1H-indol-3-yl)ethyl)acetamide, N-Acetylserotonin

M-Clarity™ Qualitätsstufe = MQ200

A1824-100MG
SIALA1824-100MGEA 203 EUR
SIALA1824-100MG
N-Acetyl-5-hydroxytryptamin, Sigma-Aldrich®
N-Acetyl-5-hydroxytryptamin
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218,3 g/mol
MDL: MFCD00005656
CAS-Nummer: 1210-83-9
EINECS: 214-916-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR