Adenosin-5'-monophosphorsäure (AMP) Monohydrat (aus Hefe) ≥97%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: A-5′-P, AMP, Adenosine-5'-monophosphoric acid monohydrate, 5′-AMP, 5′-Adenylic acid

A2252-100G A2252-25G A2252-5G
SIALA2252-100GEA 742 EUR
SIALA2252-100G SIALA2252-25G SIALA2252-5G
Adenosin-5'-monophosphorsäure (AMP) Monohydrat (aus Hefe) ≥97%, Sigma-Aldrich®
Adenosin-5'-monophosphorsäure (AMP) Monohydrat
Formel: C₁₀H₁₄N₅O₇P·1H₂O
Molecular Weight: 365.24 g/mol
MDL: MFCD00149360
CAS-Nummer: 18422-05-4
EINECS: 200-500-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR