4-Allyl-2,6-dimethoxyphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(3,5-Dimethoxy-4-hydroxyphenyl)propen

M-Clarity™ Qualitätsstufe = MQ100

A31601-5G
SIALA31601-5GEA 94.2 EUR
SIALA31601-5G
4-Allyl-2,6-dimethoxyphenol, Sigma-Aldrich®
4-Allyl-2,6-dimethoxyphenol
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
MDL: MFCD00008655
CAS-Nummer: 6627-88-9
EINECS: 229-600-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR