(±)-Allyl-2,3-epoxypropylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Allyl glycidyl ether

M-Clarity™ Qualitätsstufe = MQ200

A32608-100ML A32608-500ML
SIALA32608-100MLEA 34.4 EUR
SIALA32608-100ML SIALA32608-500ML
(±)-Allyl-2,3-epoxypropylether, Sigma-Aldrich®
(±)-Allyl-2,3-epoxypropylether
Formel: C₆H₁₀O₂
Molecular Weight: 114.14 g/mol
MDL: MFCD00005143
CAS-Nummer: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR