9-Aminoacridin Hydrochlorid Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-Aminoacridine hydrochloride hydrate, Acridin-9-amine hydrochloride hydrate

MQ200

A38401-25G A38401-5G
SIALA38401-25GEA 91 EUR
SIALA38401-25G SIALA38401-5G
9-Aminoacridin Hydrochlorid Monohydrat, Sigma-Aldrich®
9-Aminoacridin Hydrochlorid Monohydrat
Formel: C₁₃H₁₁ClN₂·1H₂O
Molecular Weight: 248.71 g/mol
MDL: MFCD00150071
CAS-Nummer: 52417-22-8
EINECS: 205-145-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR