(4-Aminophenyl)acetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

A42050-10G
SIALA42050-10GEA 125 EUR
SIALA42050-10G
(4-Aminophenyl)acetonitril, Sigma-Aldrich®
(4-Aminophenyl)acetonitril
Formel: C₈H₈N₂
MDL: MFCD00007912
CAS-Nummer: 3544-25-0
EINECS: 222-587-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR