4-Brom-1-naphthylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Amino-4-bromnaphthalin

M-Clarity™ Qualitätsstufe = MQ100

A43405-5G
SIALA43405-5GEA 97.5 EUR
SIALA43405-5G
4-Brom-1-naphthylamin, Sigma-Aldrich®
4-Brom-1-naphthylamin
Formel: C₁₀H₈BrN
Molecular Weight: 222.08 g/mol
MDL: MFCD00004023
CAS-Nummer: 2298-07-9
EINECS: 218-944-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR