Ethyl-3-amino-1H-pyrazol-4-carboxylat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
M-Clarity™ Qualitätsstufe = MQ100
A45009-5G
SIALA45009-5GEA
29.4
EUR
SIALA45009-5G
Ethyl-3-amino-1H-pyrazol-4-carboxylat, Sigma-Aldrich®
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Formel:
C₆H₉N₃O₂ Molecular Weight: 155.16 g/mol |
MDL:
MFCD00005238 CAS-Nummer: 6994-25-8 EINECS: 230-262-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...