6-Aminouracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-2,6-dihydroxypyrimidin, 4-Aminouracil, 6-Amino-2,4-dihydroxypyrimidin

M-Clarity™ Qualitätsstufe = MQ100

A50606-100G
SIALA50606-100GEA 65.9 EUR
SIALA50606-100G
6-Aminouracil, Sigma-Aldrich®
6-Aminouracil
Formel: C₄H₅N₃O₂
Molecular Weight: 127,10 g/mol
Schmelzpunkt: 360 °C
MDL: MFCD00006071
CAS-Nummer: 873-83-6
EINECS: 212-854-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR