2-Amino-4,6-dimethylpyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,6-Dimethylpyrimidin-2-ylamin, 4,6-Dimethylpyrimidin-2-amin

M-Clarity™ Qualitätsstufe = MQ100

A52005-25G
SIALA52005-25GEA 51.2 EUR
SIALA52005-25G
2-Amino-4,6-dimethylpyrimidin, Sigma-Aldrich®
2-Amino-4,6-dimethylpyrimidine
Formel: C₆H₉N₃
Molecular Weight: 123,16 g/mol
Schmelzpunkt: 152…155 °C
MDL: MFCD00006102
CAS-Nummer: 767-15-7
EINECS: 212-181-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR