6-Amino-1,3-dimethyluracil, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 4-Amino-1,3-Dimethyluracil
M-Clarity™ Qualitätsstufe = MQ100
A52153-100G
SIALA52153-100GEA
106
EUR
SIALA52153-100G
6-Amino-1,3-dimethyluracil, Sigma-Aldrich®
6-Amino-1,3-dimethyluracil
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,1546 g/mol |
MDL:
MFCD00006552 CAS-Nummer: 6642-31-5 EINECS: 229-662-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...