N-(2-Aminoethyl)morpholin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
A55004-100G
A55004-25G
A55004-5G
SIALA55004-100GEA
150
EUR
SIALA55004-100G
SIALA55004-25G
SIALA55004-5G
N-(2-Aminoethyl)morpholin, Sigma-Aldrich®
N-(2-Aminoethyl)morpholin
Formel:
C₆H₁₄N₂O |
MDL:
MFCD00006182 CAS-Nummer: 2038-03-1 EINECS: 218-011-6 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...