N-(2-Aminoethyl)morpholin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

A55004-100G A55004-25G A55004-5G
SIALA55004-100GEA 150 EUR
SIALA55004-100G SIALA55004-25G SIALA55004-5G
N-(2-Aminoethyl)morpholin, Sigma-Aldrich®
N-(2-Aminoethyl)morpholin
Formel: C₆H₁₄N₂O
MDL: MFCD00006182
CAS-Nummer: 2038-03-1
EINECS: 218-011-6
UN: 2923
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR