1-Amino-9-fluorenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Amino-9H-fluoren-9-one

A55799-1G
SIALA55799-1GEA 338 EUR
SIALA55799-1G
1-Amino-9-fluorenon, Sigma-Aldrich®
1-Amino-9-fluorenon
Formel: C₁₃H₉NO
Molecular Weight: 195,22 g/mol
MDL: MFCD00001158
CAS-Nummer: 6344-62-3
EINECS: 228-748-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR