1-Acetyl-5-bromindolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Bromoindolin-1-yl)ethanone

M-Clarity™ Qualitätsstufe = MQ200

A6125-1G
SIALA6125-1GEA 155 EUR
SIALA6125-1G
1-Acetyl-5-bromindolin, Sigma-Aldrich®
1-Acetyl-5-bromindolin
Formel: C₁₀H₁₀BrNO
Molecular Weight: 240,1 g/mol
MDL: MFCD00056017
CAS-Nummer: 22190-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR